Gene Transcripts

Gene Symbol
Unc5cl
Entrez Gene ID
76589
Gene Name
unc-5 homolog C (C. elegans)-like
Gene Aliases
2510009H09Rik, BB155270, Zud
Chromosome Location
Chr. 17: 48453839 - 48539709 on Build GRCm38
UniGene ID
Mm.311629
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152823.4 NP_690036.4 6, 7 1259
GenBank mRNA AK144925.1 - 6, 7 1260
AY510108.1 - 5, 6 1126
BC095959.1 - 6, 7 1237 5349225

View Full Product Details