Gene Transcripts

Gene Symbol
Polq
Entrez Gene ID
77782
Gene Name
polymerase (DNA directed), theta
Gene Aliases
A430110D14Rik
Chromosome Location
Chr. 16: 37011742 - 37095419 on Build GRCm38
UniGene ID
Mm.259114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159369.1 NP_001152841.1 9, 10 1541
NM_029977.2 NP_084253.1 14, 15 2378
GenBank mRNA AK156001.1 - 5, 6 846
AY074936.1 - 14, 15 2378
AY147862.1 - 9, 10 1541
AY147863.1 - 15, 16 2474
AY147864.1 - 10, 11 1637
BC138360.1 - 14, 15 2319 8861380
BC157900.1 - 14, 15 2319 9088092

View Full Product Details