Gene Transcripts

Gene Symbol
Zbtb48
Entrez Gene ID
100090
Gene Name
zinc finger and BTB domain containing 48
Gene Aliases
0610011D15Rik, AI327031, Hkr3
Chromosome Location
Chr. 4: 152019773 - 152027671 on Build GRCm38
UniGene ID
Mm.24661
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133879.2 NP_598640.2 2 282
GenBank mRNA AK027912.1 - 2 283
AM161454.1 - 2 230
BC005515.1 - 2 293 3493611
BY269141.1 - 2 283

View Full Product Details