Gene Transcripts

Gene Symbol
H2-M9
Entrez Gene ID
14997
Gene Name
histocompatibility 2, M region locus 9
Gene Aliases
M9
Chromosome Location
Chr. 17: 36639351 - 36642661 on Build GRCm38
UniGene ID
Mm.57004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008205.1 NP_032231.1 3 408
GenBank mRNA AF469488.1 - 3 408
AF539595.1 - 3 408
AK080256.1 - 3 432
BC106996.2 - 3 407

View Full Product Details