Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008584.3 | NP_032610.1 | 3 | 1005 | |
GenBank mRNA | AK028352.1 | - | 3 | 1006 | |
AK031152.1 | - | 3 | 962 | ||
AK142690.1 | - | 3 | 884 | ||
BC002076.1 | - | 3 | 904 | 3591924 | |
Z16406.1 | - | 3 | 883 |