Gene Transcripts

Gene Symbol
Tap2
Entrez Gene ID
21355
Gene Name
transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases
ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas
Chromosome Location
Chr. 17: 34204085 - 34216321 on Build GRCm38
UniGene ID
Mm.14814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011530.3 NP_035660.3 11 1975
GenBank mRNA AK020022.1 - 10 2051
AK079667.1 - 11 1889
AK145234.1 - 11 1934
AK151376.1 - 11 1885
AK151483.1 - 11 1885
AK151657.1 - 11 1908
AK153287.1 - 11 1885
AK159200.1 - 11 1941
AK161091.1 - 11 1891
AK169645.1 - 8 1183
BC005578.1 - 11 1875 3968225
BC051257.1 - 11 1888 6530488
BC057668.1 - 11 2262
CT010387.1 - 10 1822
KJ650229.1 - 10 1825
M90459.1 - 10 1822
U60087.1 - 10 1825
U60088.1 - 10 1825
U60089.1 - 10 1825
U60090.1 - 10 1825
U60091.1 - 10 1825
U60092.1 - 10 1822

View Full Product Details