Gene Transcripts

Gene Symbol
Cfdp1
Entrez Gene ID
23837
Gene Name
craniofacial development protein 1
Gene Aliases
AA408409, Bcnt, Bucentaur, Cfdp, cp27
Chromosome Location
Chr. 8: 111768473 - 111854310 on Build GRCm38
UniGene ID
Mm.279437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011801.1 NP_035931.1 6 832
GenBank mRNA AB010828.1 - 6 832
AK133989.1 - 6 850
AK160532.1 - 6 797
BC005589.1 - 6 813 3964401
Y08219.1 - 6 831

View Full Product Details