Gene Transcripts

Gene Symbol
Mreg
Entrez Gene ID
381269
Gene Name
melanoregulin
Gene Aliases
Gm974, Wdt2, dsu
Chromosome Location
Chr. 1: 72159233 - 72212307 on Build GRCm38
UniGene ID
Mm.422636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005423.2 NP_001005423.1 4 625
GenBank mRNA AY628210.1 - 4 429
BC068125.1 - 4 625 6828150

View Full Product Details