Gene Transcripts

Gene Symbol
Srpk3
Entrez Gene ID
56504
Gene Name
serine/arginine-rich protein specific kinase 3
Gene Aliases
Mssk1, Stk23
Chromosome Location
Chr. X: 73773261 - 73778925 on Build GRCm38
UniGene ID
Mm.111904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019684.1 NP_062658.1 6, 7 614
GenBank mRNA AF043288.1 - 6, 7 614
AJ748651.1 - 5, 6 438
BC120883.1 - 6, 7 609 40057934
BC120884.1 - 6, 7 609 40057937

View Full Product Details