Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001047435.2 | NP_001040900.1 | 3 | 1108 | |
NM_001047436.2 | NP_001040901.1 | 3 | 1124 | ||
NM_001286344.1 | NP_001273273.1 | 3 | 1093 | ||
NM_001286345.1 | NP_001273274.1 | 3 | 1021 | ||
NM_019695.3 | NP_062669.2 | 3 | 1111 | ||
GenBank mRNA | AF070970.1 | - | 3 | 1107 | |
AK003030.1 | - | 3 | 1077 | ||
AK158848.1 | - | 3 | 1106 | ||
BC049593.1 | - | 3 | 1085 | 6771847 | |
BC061114.1 | - | 3 | 1021 | 6535523 | |
BF466814.1 | - | 1 | 169 | ||
CO040458.1 | - | 1 | 170 |