Gene Transcripts

Gene Symbol
Lat2
Entrez Gene ID
56743
Gene Name
linker for activation of T cells family, member 2
Gene Aliases
AW125574, LAB, NTAL, WSCR5, Wbscr15, Wbscr5
Chromosome Location
Chr. 5: 134600103 - 134619783 on Build GRCm38
UniGene ID
Mm.391375
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020044.3 NP_064428.1 12 732
NM_022964.4 NP_075253.2 11 696
GenBank mRNA AF257136.1 - 11 676
AK134721.1 - 9 424
AK138953.1 - 12 691
AK143533.1 - 11 697
AK144261.1 - 12 671
AK162321.1 - 11 655
AY190024.1 - 12 697
BC005804.1 - 11 625 3154073

View Full Product Details