Gene Transcripts

Gene Symbol
Retnla
Entrez Gene ID
57262
Gene Name
resistin like alpha
Gene Aliases
1810019L16Rik, Fizz-1, Fizz1, HIMF, RELM-alpha, RELMa, RELMalpha, Xcp2
Chromosome Location
Chr. 16: 48842552 - 48844461 on Build GRCm38
UniGene ID
Mm.441868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020509.3 NP_065255.2 4 328
GenBank mRNA AF205951.1 - 3 261
AF290872.1 - 4 366
AF316397.2 - 4 270
AF323082.1 - 4 338
AK007553.1 - 4 273
BC029248.1 - 3 268 4189285

View Full Product Details