Gene Transcripts

Gene Symbol
Gsdma
Entrez Gene ID
57911
Gene Name
gasdermin A
Gene Aliases
BB149167, Gsdm, Gsdm1, Gsdma1, H312E
Chromosome Location
Chr. 11: 98664337 - 98677708 on Build GRCm38
UniGene ID
Mm.458085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021347.4 NP_067322.1 7, 8 815
GenBank mRNA AB033595.1 - 7, 8 804
AK028645.1 - 7, 8 816
AK028698.1 - 7, 8 816
AK029062.1 - 7, 8 799

View Full Product Details