Gene Transcripts

Gene Symbol
Unc50
Entrez Gene ID
67387
Gene Name
unc-50 homolog (C. elegans)
Gene Aliases
1110002A21Rik, GMH1, UNCL, URP
Chromosome Location
Chr. 1: 37430144 - 37439124 on Build GRCm38
UniGene ID
Mm.27404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026123.3 NP_080399.1 2 364
GenBank mRNA AK003273.1 - 2 139
AK004433.1 - 2 128
AK132757.1 - 2 365
AK135885.1 - 2 356
AK217920.1 - 2 85
AY017214.1 - 2 349
BC019484.1 - 2 331 4039903

View Full Product Details