Gene Transcripts

Gene Symbol
Kctd4
Entrez Gene ID
67516
Gene Name
potassium channel tetramerisation domain containing 4
Gene Aliases
2210017A09Rik, AU017169
Chromosome Location
Chr. 14: 75955003 - 75965217 on Build GRCm38
UniGene ID
Mm.390712
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026214.3 NP_080490.3 2 1124
GenBank mRNA AK017592.1 - 1 855
AK032314.1 - 1 831
AK032624.1 - 1 862
BC028748.1 - 1 839 1332429

View Full Product Details