Gene Transcripts

Gene Symbol
Klhl40
Entrez Gene ID
72330
Gene Name
kelch-like 40
Gene Aliases
2310024D23Rik, Kbtbd5
Chromosome Location
Chr. 9: 121777101 - 121791002 on Build GRCm38
UniGene ID
Mm.45734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028202.3 NP_082478.1 1 1225
GenBank mRNA AK009491.1 - 1 1141
AK078669.1 - 1 1139
AK160799.1 - 1 1085
BC125576.1 - 1 1225 40130091
BC137889.1 - 1 1225 8860908

View Full Product Details