Gene Transcripts

Gene Symbol
Dbr1
Entrez Gene ID
83703
Gene Name
debranching enzyme homolog 1 (S. cerevisiae)
Gene Aliases
AW018415
Chromosome Location
Chr. 9: 99575775 - 99584461 on Build GRCm38
UniGene ID
Mm.31324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031403.3 NP_113580.2 2 387
GenBank mRNA AK050090.1 - 2 388
AK090351.1 - 2 363
AK139631.1 - 2 369
BC006661.1 - 2 382 3501390

View Full Product Details