Gene Transcripts

Gene Symbol
Pid1
Entrez Gene ID
98496
Gene Name
phosphotyrosine interaction domain containing 1
Gene Aliases
5033414K04Rik, NYGGF4
Chromosome Location
Chr. 1: 84036293 - 84284675 on Build GRCm38
UniGene ID
Mm.187470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003948.2 NP_001003948.2 3 660
GenBank mRNA AK035542.1 - 3 535
AK083256.1 - 3 581
AK150971.1 - 3 598
AK151267.1 - 3 630
BC054112.1 - 2 293
BC080290.1 - 3 606 6816865

View Full Product Details