Gene Transcripts

Gene Symbol
Trip10
Entrez Gene ID
106628
Gene Name
thyroid hormone receptor interactor 10
Gene Aliases
AI646975, Cip4
Chromosome Location
Chr. 17: 57233207 - 57263697 on Build GRCm38
UniGene ID
Mm.37368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242389.1 NP_001229318.1 8, 9 895
NM_001242390.1 NP_001229319.1 8, 9 895
NM_001242391.1 NP_001229320.1 8, 9 895
NM_134125.4 NP_598886.1 8, 9 895
GenBank mRNA AF502565.1 - 8, 9 798
AK088909.1 - 8, 9 845
AK149902.1 - 8, 9 867
AY081142.1 - 8, 9 846
BC003249.1 - 8, 9 897 3500118

View Full Product Details