Gene Transcripts

Gene Symbol
Nkx1-2
Entrez Gene ID
20231
Gene Name
NK1 transcription factor related, locus 2 (Drosophila)
Gene Aliases
Nkx-1.1, Nkx-1.2, Sax1
Chromosome Location
Chr. 7: 132596239 - 132599637 on Build GRCm38
UniGene ID
Mm.334281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009123.2 NP_033149.1 2 1015
GenBank mRNA AF222444.1 - 2 1015
BC139757.1 - 2 944 40047346
X75384.1 - 2 1120

View Full Product Details