Gene Transcripts

Gene Symbol
Spink10
Entrez Gene ID
328971
Gene Name
serine peptidase inhibitor, Kazal type 10
Gene Aliases
A230091H23Rik
Chromosome Location
Chr. 18: 62548911 - 62662933 on Build GRCm38
UniGene ID
Mm.312675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177829.3 NP_808497.1 8 610
GenBank mRNA AK039062.1 - 8 611
AK162840.1 - 8 574
AV380905.2 - 3 146
BC049733.1 - 267
BC125360.1 - 8 526 40129875
BC125362.1 - 8 526 40129877
DQ437330.1 - 8 610

View Full Product Details