Gene Transcripts

Gene Symbol
Msl3l2
Entrez Gene ID
73390
Gene Name
male-specific lethal 3-like 2 (Drosophila)
Gene Aliases
1700060H10Rik
Chromosome Location
Chr. 10: 56083827 - 56116880 on Build GRCm38
UniGene ID
Mm.181475
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163833.1 NP_001157305.1 2 515
GenBank mRNA AK155382.1 - 2 501
BC013191.1 - 1 117
BC037487.1 - 2 526

View Full Product Details