Gene Transcripts

Gene Symbol
Sp6
Entrez Gene ID
83395
Gene Name
trans-acting transcription factor 6
Gene Aliases
1110025J03Rik, AA591031, AI592962, Epfn, Klf14
Chromosome Location
Chr. 11: 97013569 - 97024739 on Build GRCm38
UniGene ID
Mm.156282
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031183.2 NP_112460.1 2 401
GenBank mRNA AK137686.1 - 2 625
AM295492.1 - 2 645
AY338955.1 - 2 401
BC099583.1 - 1 343
BC138818.1 - 1 347 8861840
BC138822.1 - 1 347 8861844
BE648643.1 - 1 18

View Full Product Details