Gene Transcripts

Gene Symbol
Hepacam2
Entrez Gene ID
101202
Gene Name
HEPACAM family member 2
Gene Aliases
AI987662
Chromosome Location
Chr. 6: 3457089 - 3498189 on Build GRCm38
UniGene ID
Mm.206911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178899.5 NP_849230.1 3 708
GenBank mRNA AK033728.1 - 3 710
AK050501.1 - 3 709
BC096374.1 - 3 695 4483627

View Full Product Details