Gene Transcripts
- Gene Symbol
- Csn1s2a
- Entrez Gene ID
- 12993
- Gene Name
- casein alpha s2-like A
- Gene Aliases
- Csng, PP22
- Chromosome Location
- Chr. 5: 87774567 - 87788797 on Build GRCm38
- UniGene ID
- Mm.474059
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142688.1 |
- |
4, 5 |
162 |
|
D10215.1 |
- |
4, 5 |
167 |
|
View Full Product Details