Gene Transcripts

Gene Symbol
Hc
Entrez Gene ID
15139
Gene Name
hemolytic complement
Gene Aliases
C5, C5a, He
Chromosome Location
Chr. 2: 34983331 - 35061441 on Build GRCm38
UniGene ID
Mm.2168
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010406.2 NP_034536.1 5, 6 650
GenBank mRNA AK080166.1 - 5, 6 649
M35525.1 - 5, 6 605
M35526.1 - 5, 6 605

View Full Product Details