Gene Transcripts

Gene Symbol
Hoxd8
Entrez Gene ID
15437
Gene Name
homeobox D8
Gene Aliases
4921540P06Rik, AI047735, Hox-4.3, Hox-5.4
Chromosome Location
Chr. 2: 74704615 - 74707933 on Build GRCm38
UniGene ID
Mm.280673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290731.1 NP_001277660.1 2, 3 269
NM_008276.3 NP_032302.2 1, 2 641
GenBank mRNA AK016033.1 - 1, 2 370
BX513411.1 - 2, 3 293

View Full Product Details