Gene Transcripts

Gene Symbol
Fabp6
Entrez Gene ID
16204
Gene Name
fatty acid binding protein 6, ileal (gastrotropin)
Gene Aliases
GT, I-15P, I-BABP, ILBP, ILBP3, Illbp
Chromosome Location
Chr. 11: 43596040 - 43601562 on Build GRCm38
UniGene ID
Mm.142716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008375.2 NP_032401.1 1 66
GenBank mRNA BC119289.1 - 1 36
BC120767.1 - 1 36 40129690
CJ043022.1 - 1 66

View Full Product Details