Gene Transcripts

Gene Symbol
Prima1
Entrez Gene ID
170952
Gene Name
proline rich membrane anchor 1
Gene Aliases
B230212M13Rik, BB120497, PRiMA
Chromosome Location
Chr. 12: 103194453 - 103242720 on Build GRCm38
UniGene ID
Mm.217019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133364.2 NP_579942.2 3 228
GenBank mRNA AK140034.1 - 3 229
AY043275.1 - 3 229
AY225515.1 - 3 229

View Full Product Details