Gene Transcripts

Gene Symbol
Msx1
Entrez Gene ID
17701
Gene Name
msh homeobox 1
Gene Aliases
AA675338, AI324650, Hox-7, Hox7, Hox7.1, msh
Chromosome Location
Chr. 5: 37820491 - 37824585 on Build GRCm38
UniGene ID
Mm.256509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010835.2 NP_034965.2 1 624
GenBank mRNA AF308572.1 - 1 408
AK081534.1 - 1 624
AK145726.1 - 1 622
BC016426.1 - 1 607 4923403
BU922533.1 - 1 165
X14759.1 - 1 495
X59251.1 - 1 495

View Full Product Details