Gene Transcripts

Gene Symbol
Myl4
Entrez Gene ID
17896
Gene Name
myosin, light polypeptide 4
Gene Aliases
ALC1, AMLC, ELC, ELC1a, GT1, MLC1a, Myla
Chromosome Location
Chr. 11: 104550530 - 104587219 on Build GRCm38
UniGene ID
Mm.390355
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010858.4 NP_034988.2 5 338
GenBank mRNA AK013098.1 - 3 266
AK084544.1 - 1 1976
BC048485.1 - 5 338 6741975
M19436.1 - 1 20

View Full Product Details