Gene Transcripts

Gene Symbol
Neurog1
Entrez Gene ID
18014
Gene Name
neurogenin 1
Gene Aliases
AKA, Math4C, Neurod3, bHLHa6, ngn1
Chromosome Location
Chr. 13: 56250492 - 56252163 on Build GRCm38
UniGene ID
Mm.266665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010896.2 NP_035026.1 1 260
GenBank mRNA BC062148.1 - 1 260

View Full Product Details