Gene Transcripts

Gene Symbol
Nkx2-2
Entrez Gene ID
18088
Gene Name
NK2 homeobox 2
Gene Aliases
Nkx-2.2, Nkx2.2, tinman
Chromosome Location
Chr. 2: 147177546 - 147195783 on Build GRCm38
UniGene ID
Mm.330639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077632.1 NP_001071100.1 2 716
NM_010919.2 NP_035049.1 2 716
GenBank mRNA AK043561.1 - 2 570
AK141002.1 - 2 721
BC138159.1 - 3 537 8861178
BC138160.1 - 3 537 8861179
U31566.1 - 2 675

View Full Product Details