Gene Transcripts

Gene Symbol
Pvrl2
Entrez Gene ID
19294
Gene Name
poliovirus receptor-related 2
Gene Aliases
AI325026, AI987993, Cd112, MPH, Pvr, Pvs, nectin-2
Chromosome Location
Chr. 7: 19716644 - 19749573 on Build GRCm38
UniGene ID
Mm.4341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159724.1 NP_001153196.1 3 945
NM_008990.3 NP_033016.3 3 945
GenBank mRNA AK075815.1 - 3 850
AK157513.1 - 3 850
BC009088.1 - 3 838 2812478
BC046770.1 - 3 841 5703547
BC059941.1 - 3 897 6530953
D26107.1 - 3 674
M80206.1 - 3 674

View Full Product Details