Gene Transcripts

Gene Symbol
Rab5c
Entrez Gene ID
19345
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
AI326010, Rabl
Chromosome Location
Chr. 11: 100715009 - 100738210 on Build GRCm38
UniGene ID
Mm.29829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB232595.1 - 1 53
AK089199.1 - 2 255
AK134691.1 - 2 194
AK159832.1 - 2 244
AK167599.1 - 2 244
AK170540.1 - 2 264
AK172328.1 - 2 201
BC023027.1 - 2 210 5369566
BC027378.1 - 1 16
BC029678.1 - 2 194 5368061
BY081527.1 - 2 279

View Full Product Details