Gene Transcripts

Gene Symbol
Sept8
Entrez Gene ID
20362
Gene Name
septin 8
Gene Aliases
AW046166, Sepl, mKIAA0202
Chromosome Location
Chr. 11: 53519736 - 53549600 on Build GRCm38
UniGene ID
Mm.274399
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252332.1 NP_001239261.1 5 853
NM_001252333.1 NP_001239262.1 5 847
NM_033144.2 NP_149156.1 5 853
GenBank mRNA AB093215.1 - 5 703
AF179996.1 - 4 549
AK144178.1 - 5 689
BC037707.1 - 5 641
BC049819.1 - 5 668
BC059248.1 - 5 789 6400429

View Full Product Details