Gene Transcripts
- Gene Symbol
- Snrnp70
- Entrez Gene ID
- 20637
- Gene Name
- small nuclear ribonucleoprotein 70 (U1)
- Gene Aliases
- 2700022N21Rik, 3200002N22Rik, AI325098, R74807, Rnu1p70, Rnulp70, Snrp70, Srnp70, U1-70, U1-70K
- Chromosome Location
- Chr. 7: 45376454 - 45395742 on Build GRCm38
- UniGene ID
- Mm.216386
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005045.2 |
- |
6, 7 |
594 |
|
AK011564.1 |
- |
6, 7 |
550 |
|
AK012272.1 |
- |
4, 5 |
243 |
|
AK014311.2 |
- |
6, 7 |
591 |
|
AK077425.1 |
- |
6, 7 |
596 |
|
AK133115.1 |
- |
6, 7 |
595 |
|
AK146729.1 |
- |
6, 7 |
595 |
|
AK148013.1 |
- |
6, 7 |
621 |
|
BC049128.1 |
- |
6, 7 |
560 |
|
BC051414.1 |
- |
5, 6 |
395 |
|
BC131994.1 |
- |
6, 7 |
487 |
40130271 |
BC132020.1 |
- |
6, 7 |
487 |
40130297 |
View Full Product Details