Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011458.2 | NP_035588.2 | 3, 4 | 998 | |
GenBank mRNA | AK004842.1 | - | 3, 4 | 995 | |
AK149436.1 | - | 3, 4 | 999 | ||
BC011217.1 | - | 3, 4 | 975 | 4195356 | |
BC016407.1 | - | 3, 4 | 986 | 4210846 | |
BC019802.1 | - | 3, 4 | 975 | 5050045 | |
D00725.1 | - | 976 | |||
X55147.1 | - | 3, 4 | 981 | ||
X56786.1 | - | 3, 4 | 987 |