Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037321.1 | NP_001032398.1 | 3 | 2475 | |
GenBank mRNA | AK036385.1 | - | 4 | 2233 | |
AK220373.1 | - | 2 | 2109 | ||
BC107406.1 | - | 3 | 2475 | 40058923 | |
BC107407.1 | - | 2 | 444 |