Gene Transcripts

Gene Symbol
Ism2
Entrez Gene ID
217738
Gene Name
isthmin 2 homolog (zebrafish)
Gene Aliases
Gm264, Thsd3
Chromosome Location
Chr. 12: 87278638 - 87300346 on Build GRCm38
UniGene ID
Mm.219363
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290302.1 NP_001277231.1 3 473
GenBank mRNA BU054611.1 - 2 278

View Full Product Details