Gene Transcripts

Gene Symbol
Tnnc2
Entrez Gene ID
21925
Gene Name
troponin C2, fast
Gene Aliases
Tncs
Chromosome Location
Chr. 2: 164777161 - 164779734 on Build GRCm38
UniGene ID
Mm.1716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009394.2 NP_033420.1 5 455
GenBank mRNA AK133548.1 - 5 451
BC024390.1 - 5 454 5026329
CB566378.1 - 490

View Full Product Details