Gene Transcripts

Gene Symbol
D17Wsu92e
Entrez Gene ID
224647
Gene Name
DNA segment, Chr 17, Wayne State University 92, expressed
Gene Aliases
AU020189, C030010A15, C80239
Chromosome Location
Chr. 17: 27751232 - 27820542 on Build GRCm38
UniGene ID
Mm.196577
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033279.3 NP_001028451.2 1, 2 371
NM_001044719.2 NP_001038184.1 1, 2 371
NM_001271511.1 NP_001258440.1 1, 2 371
GenBank mRNA AK143016.1 - 1, 2 284
AK161603.1 - 1, 2 372
BC132324.1 - 2, 3 297 40130601
BC132326.1 - 2, 3 297 40130603
BC137573.1 - 1, 2 371 8860592
BC137576.1 - 1, 2 371 8860595
BC145403.1 - 1, 2 371 9053961
BY755073.1 - 1, 2 297
CX220817.1 - 1, 2 146

View Full Product Details