Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177663.4 | NP_808331.1 | 2 | 866 | |
GenBank mRNA | AK053778.1 | - | 2 | 867 | |
AK076747.1 | - | 2 | 588 | ||
AK155979.1 | - | 2 | 870 | ||
AK178586.1 | - | 1 | 158 | ||
BC098326.1 | - | 2 | 853 | 5720443 |