Gene Transcripts

Gene Symbol
Vps13d
Entrez Gene ID
230895
Gene Name
vacuolar protein sorting 13 D (yeast)
Gene Aliases
Chromosome Location
Chr. 4: 144871045 - 145195095 on Build GRCm38
UniGene ID
Mm.240310
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276465.1 NP_001263394.1 49 10088
NM_001276502.1 NP_001263431.1 48 10004
GenBank mRNA AK172952.1 - 18 2516
BC037490.1 - 4 596

View Full Product Details