Gene Transcripts

Gene Symbol
Slc45a1
Entrez Gene ID
242773
Gene Name
solute carrier family 45, member 1
Gene Aliases
C230078B22, Dnb5
Chromosome Location
Chr. 4: 150629396 - 150652174 on Build GRCm38
UniGene ID
Mm.29280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173774.3 NP_776135.2 6 1719
GenBank mRNA AK082651.1 - 1712
AK134677.1 - 6 1645
AK134683.1 - 6 1710
BC051188.1 - 6 1681

View Full Product Details