Gene Transcripts

Gene Symbol
Svs1
Entrez Gene ID
243377
Gene Name
seminal vesicle secretory protein 1
Gene Aliases
9530003A11
Chromosome Location
Chr. 6: 48986861 - 48991725 on Build GRCm38
UniGene ID
Mm.446302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172888.3 NP_766476.2 1 383
GenBank mRNA AK035222.1 - 1 384
AY115568.1 - 1 340
AY283179.1 - 1 340
BC125635.1 - 1 254 40130150

View Full Product Details