Gene Transcripts

Gene Symbol
Vmn2r24
Entrez Gene ID
243628
Gene Name
vomeronasal 2, receptor 24
Gene Aliases
EG243628
Chromosome Location
Chr. 6: 123778971 - 123816280 on Build GRCm38
UniGene ID
Mm.469873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104639.1 NP_001098109.1 1 153

View Full Product Details