Gene Transcripts

Gene Symbol
Ggn
Entrez Gene ID
243897
Gene Name
gametogenetin
Gene Aliases
AI593290
Chromosome Location
Chr. 7: 29170210 - 29173939 on Build GRCm38
UniGene ID
Mm.389886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182694.2 NP_874353.2 3 2531
NM_182696.2 NP_874355.1 3 487
GenBank mRNA AF538032.1 - 3 2020
AF538033.1 - 4 1440
AF538034.1 - 3 487
BC089358.1 - 3 2531 30610755

View Full Product Details