Gene Transcripts

Gene Symbol
Gin1
Entrez Gene ID
252876
Gene Name
gypsy retrotransposon integrase 1
Gene Aliases
4930429M06Rik, 8430411H10Rik, Gin-1, Zh2c2
Chromosome Location
Chr. 1: 97770168 - 97793710 on Build GRCm38
UniGene ID
Mm.86090
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026250.2 NP_080526.2 2 196
GenBank mRNA AK014287.1 - 2 208
AK015243.1 - 2 175
AK018404.1 - 2 197
AK084620.1 - 1 100
AK090002.1 - 2 181
AK169361.1 - 2 159
BC033572.1 - 1 141 5067692

View Full Product Details