Gene Transcripts

Gene Symbol
Mapk10
Entrez Gene ID
26414
Gene Name
mitogen-activated protein kinase 10
Gene Aliases
C230008H04Rik, JNK3, JNK3B1, JNK3B2, SAPK(beta), Serk2, p493F12, p54bSAPK
Chromosome Location
Chr. 5: 102907948 - 103211339 on Build GRCm38
UniGene ID
Mm.39253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081567.1 NP_001075036.1 11 1572
NM_009158.2 NP_033184.2 11 1572
GenBank mRNA AB005665.1 - 8 955
AB096076.1 - 9 1006
AB096077.1 - 9 1006
AB096078.1 - 9 1006
AB096079.1 - 9 1006
AK036275.1 - 11 1524
AK042368.1 - 10 1137
AK163206.1 - 11 1523
BC046625.1 - 11 1511 6494163
L35236.1 - 11 1315

View Full Product Details